Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC6
Entrez Gene ID
319089
Gene Name
tetratricopeptide repeat domain 6
Gene Aliases
C14orf25, NCRNA00291
Chromosome Location
Chr.14: 37595199 - 37842628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001310135.1 NP_001297064.1 27 - 28 5591 66
XM_011537430.2 XP_011535732.1 23 - 24 3423 66
XM_011537431.2 XP_011535733.1 21 - 22 3080 66
XM_017021254.1 XP_016876743.1 26 - 27 5441 66
XM_017021255.1 XP_016876744.1 25 - 26 5306 66
XM_017021256.1 XP_016876745.1 26 - 27 5705 66
GenBank mRNA BC103914.2 - 8 - 9 940 66
BC103915.1 - 8 - 9 940 66
BC103916.2 - 8 - 9 940 66
BX161415.1 - 7 - 8 875 66

View Full Product Details