Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD84
Entrez Gene ID
8832
Gene Name
CD84 molecule
Gene Aliases
LY9B, SLAMF5, hCD84, mCD84
Chromosome Location
Chr.1: 160541094 - 160579516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184879.1 NP_001171808.1 2 - 3 471 79
NM_001184881.1 NP_001171810.1 2 - 3 471 79
NM_003874.3 NP_003865.1 2 - 3 471 79
XM_011510094.2 XP_011508396.1 2 - 3 452 79
GenBank mRNA AF054815.1 - 2 - 3 461 79
AF054816.1 - 2 - 3 461 79
AF054817.1 - 2 - 3 461 79
AF054818.1 - 2 - 3 461 79
AJ223324.1 - 2 - 3 436 79
AK313496.1 - 2 - 3 437 79
BC020063.1 - 2 - 3 416 79
CR541847.1 - 2 - 3 392 79
U82988.1 - 2 - 3 433 79
U96627.1 - 2 - 3 461 79
Y12632.1 - 2 - 3 461 79

View Full Product Details