The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024316.2 | NP_077292.2 | 1 - 2 | 184 | 87 |
GenBank mRNA | BC014986.1 | - | 1 - 2 | 149 | 87 |
BP870984.1 | - | 1 - 2 | 185 | 87 |