The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017666.4 | NP_060136.1 | 14 - 15 | 2046 | 91 |
XM_006724765.3 | XP_006724828.1 | 13 - 14 | 1850 | 91 | |
GenBank mRNA | AK290373.1 | - | 14 - 15 | 1997 | 91 |
AL133204.1 | - | 13 - 14 | 1834 | 91 | |
AL834333.1 | - | 14 - 15 | 91 | ||
BC051728.1 | - | 14 - 15 | 91 | ||
DA441065.1 | - | 1 - 2 | 70 | 91 |