The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017666.4 | NP_060136.1 | 6 - 7 | 691 | 131 |
XM_006724765.3 | XP_006724828.1 | 6 - 7 | 642 | 131 | |
GenBank mRNA | AK000102.1 | - | 6 - 7 | 614 | 131 |
AK290373.1 | - | 6 - 7 | 642 | 131 | |
AL133204.1 | - | 6 - 7 | 626 | 131 | |
AL834333.1 | - | 6 - 7 | 622 | 131 | |
BC051728.1 | - | 6 - 7 | 642 | 131 |