Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBX10
Entrez Gene ID
347853
Gene Name
T-box 10
Gene Aliases
TBX13, TBX7
Chromosome Location
Chr.11: 67631303 - 67639560 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005995.4 NP_005986.2 5 - 6 789 78
XM_011544992.2 XP_011543294.1 4 - 5 1079 78
XM_011544993.1 XP_011543295.1 3 - 4 415 78
GenBank mRNA AY229977.1 - 5 - 6 790 78
BC113485.1 - 5 - 6 783 78
BC113487.1 - 5 - 6 783 78

View Full Product Details