Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRR12
Entrez Gene ID
57479
Gene Name
proline rich 12
Gene Aliases
KIAA1205
Chromosome Location
Chr.19: 49591177 - 49626439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB033031.1 - 6 - 7 2566 89
BC034003.1 - 4 - 5 932 89

View Full Product Details