Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC39A10
Entrez Gene ID
57181
Gene Name
solute carrier family 39 member 10
Gene Aliases
LZT-Hs2
Chromosome Location
Chr.2: 195575949 - 195737702 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127257.1 NP_001120729.1 4 - 5 1599 67
NM_020342.2 NP_065075.1 4 - 5 1550 67
XM_005246689.4 XP_005246746.2 4 - 5 1557 67
XM_011511504.2 XP_011509806.1 4 - 5 1464 67
XM_011511505.2 XP_011509807.1 5 - 6 1628 67
XM_011511506.2 XP_011509808.1 4 - 5 4287 67
XM_011511507.2 XP_011509809.1 4 - 5 1557 67
XM_017004522.1 XP_016860011.1 4 - 5 2589 67
GenBank mRNA AB033091.1 - 4 - 5 1463 67
AK291241.1 - 4 - 5 1462 67
AK294771.1 - 5 - 6 646 67
AL050294.1 - 4 - 5 1449 67
BC101516.1 - 4 - 5 1459 67
BC112223.1 - 4 - 5 1471 67
CR749813.1 - 4 - 5 1450 67

View Full Product Details