Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC39A10
Entrez Gene ID
57181
Gene Name
solute carrier family 39 member 10
Gene Aliases
LZT-Hs2
Chromosome Location
Chr.2: 195575949 - 195737702 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127257.1 NP_001120729.1 5 - 6 1790 148
NM_020342.2 NP_065075.1 5 - 6 1741 148
XM_005246689.4 XP_005246746.2 5 - 6 1748 148
XM_011511504.2 XP_011509806.1 5 - 6 1655 148
XM_011511505.2 XP_011509807.1 6 - 7 1819 148
XM_011511506.2 XP_011509808.1 5 - 6 4478 148
XM_011511507.2 XP_011509809.1 5 - 6 1748 148
XM_017004522.1 XP_016860011.1 5 - 6 2780 148
GenBank mRNA AB033091.1 - 5 - 6 1654 148
AK291241.1 - 5 - 6 1653 148
AK294771.1 - 6 - 7 837 148
BC073909.1 - 1 - 2 166 148
BC101516.1 - 5 - 6 1650 148
BC112223.1 - 5 - 6 1662 148
CR749813.1 - 5 - 6 1641 148

View Full Product Details