Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPN1
Entrez Gene ID
11321
Gene Name
GPN-loop GTPase 1
Gene Aliases
ATPBD1A, MBDIN, NTPBP, RPAP4, XAB1
Chromosome Location
Chr.2: 27628648 - 27650846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145047.1 NP_001138519.1 4 - 5 265 97
NM_001145048.1 NP_001138520.1 3 - 4 197 97
NM_007266.3 NP_009197.2 3 - 4 304 97
GenBank mRNA AB044661.1 - 3 - 4 265 97
AJ010842.1 - 3 - 4 194 97
AK298827.1 - 3 - 4 598 97
AK298866.1 - 3 - 4 304 97
AK310340.1 - 4 - 5 255 97
AK312992.1 - 3 - 4 276 97
BC007451.2 - 3 - 4 248 97

View Full Product Details