The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243571.1 | NP_001230500.1 | 11 - 12 | 1321 | 79 |
NM_016156.5 | NP_057240.3 | 8 - 9 | 1054 | 79 | |
NM_201278.2 | NP_958435.1 | 10 - 11 | 1248 | 79 | |
NM_201281.2 | NP_958438.1 | 9 - 10 | 1125 | 79 | |
XM_005274374.2 | XP_005274431.1 | 9 - 10 | 966 | 79 | |
XM_005274375.2 | XP_005274432.1 | 9 - 10 | 1080 | 79 | |
XM_006718934.2 | XP_006718997.1 | 10 - 11 | 1160 | 79 | |
XM_006718935.2 | XP_006718998.1 | 10 - 11 | 1090 | 79 | |
XM_006718936.3 | XP_006718999.1 | 9 - 10 | 1092 | 79 | |
XM_011543058.2 | XP_011541360.1 | 11 - 12 | 1182 | 79 | |
XM_011543059.2 | XP_011541361.1 | 11 - 12 | 1016 | 79 | |
XM_017018517.1 | XP_016874006.1 | 10 - 11 | 1187 | 79 | |
XM_017018518.1 | XP_016874007.1 | 9 - 10 | 969 | 79 | |
GenBank mRNA | AB028996.1 | - | 8 - 9 | 1142 | 79 |
AK291277.1 | - | 8 - 9 | 828 | 79 | |
AK293243.1 | - | 10 - 11 | 959 | 79 | |
AK302940.1 | - | 9 - 10 | 851 | 79 | |
BC034474.1 | - | 10 - 11 | 1145 | 79 | |
BC040432.1 | - | 8 - 9 | 806 | 79 | |
BC052990.1 | - | 8 - 9 | 939 | 79 | |
BX647997.1 | - | 9 - 10 | 1019 | 79 |