The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134937.1 | NP_001128409.1 | 16 - 17 | 1670 | 87 |
NM_001134938.1 | NP_001128410.1 | 13 - 14 | 1466 | 87 | |
NM_030917.3 | NP_112179.2 | 16 - 17 | 1688 | 87 | |
XM_005265769.4 | XP_005265826.1 | 17 - 18 | 1782 | 87 | |
XM_005265773.4 | XP_005265830.1 | 16 - 17 | 1715 | 87 | |
XM_005265774.4 | XP_005265831.1 | 15 - 16 | 1627 | 87 | |
XM_005265781.4 | XP_005265838.1 | 15 - 16 | 1599 | 87 | |
XM_005265782.4 | XP_005265839.1 | 14 - 15 | 1518 | 87 | |
XM_017008662.1 | XP_016864151.1 | 15 - 16 | 1660 | 87 | |
XM_017008663.1 | XP_016864152.1 | 16 - 17 | 1616 | 87 | |
XM_017008664.1 | XP_016864153.1 | 15 - 16 | 1671 | 87 | |
XM_017008665.1 | XP_016864154.1 | 15 - 16 | 1587 | 87 | |
XM_017008666.1 | XP_016864155.1 | 14 - 15 | 1604 | 87 | |
XM_017008667.1 | XP_016864156.1 | 15 - 16 | 1570 | 87 | |
XM_017008668.1 | XP_016864157.1 | 14 - 15 | 1616 | 87 | |
XM_017008669.1 | XP_016864158.1 | 14 - 15 | 1516 | 87 | |
GenBank mRNA | AK123992.1 | - | 7 - 8 | 2186 | 87 |
AK295737.1 | - | 16 - 17 | 1685 | 87 | |
AL136910.1 | - | 13 - 14 | 1447 | 87 | |
AY366510.1 | - | 16 - 17 | 1645 | 87 | |
BC011543.1 | - | 7 - 8 | 706 | 87 | |
BC024016.1 | - | 14 - 15 | 2730 | 87 | |
BC026724.1 | - | 5 - 6 | 510 | 87 | |
BC052959.1 | - | 14 - 15 | 2725 | 87 | |
BC110383.1 | - | 16 - 17 | 1681 | 87 |