Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MICAL3
Entrez Gene ID
57553
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 3
Gene Aliases
MICAL-3
Chromosome Location
Chr.22: 17787649 - 18024559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122731.2 NP_001116203.1 10 - 11 1623 87
NM_001136004.3 NP_001129476.1 10 - 11 1623 87
NM_015241.2 NP_056056.2 11 - 12 1902 87
XM_005261262.3 XP_005261319.1 11 - 12 1732 87
XM_011546137.2 XP_011544439.1 11 - 12 1732 87
XM_011546139.2 XP_011544441.1 11 - 12 1732 87
XM_011546142.2 XP_011544444.1 11 - 12 1732 87
XM_011546146.2 XP_011544448.1 11 - 12 1732 87
XM_017028865.1 XP_016884354.1 11 - 12 2250 87
XM_017028866.1 XP_016884355.1 11 - 12 2056 87
XM_017028867.1 XP_016884356.1 11 - 12 3173 87
XM_017028868.1 XP_016884357.1 11 - 12 1902 87
XM_017028869.1 XP_016884358.1 11 - 12 1647 87
XM_017028870.1 XP_016884359.1 11 - 12 1648 87
XM_017028871.1 XP_016884360.1 11 - 12 1732 87
XM_017028872.1 XP_016884361.1 11 - 12 1732 87
XM_017028873.1 XP_016884362.1 11 - 12 1732 87
XM_017028874.1 XP_016884363.1 11 - 12 1732 87
XM_017028875.1 XP_016884364.1 11 - 12 1732 87
XM_017028876.1 XP_016884365.1 11 - 12 1732 87
XM_017028877.1 XP_016884366.1 11 - 12 1732 87
XM_017028878.1 XP_016884367.1 11 - 12 1732 87
XM_017028879.1 XP_016884368.1 11 - 12 1732 87
XM_017028880.1 XP_016884369.1 11 - 12 2055 87
XM_017028881.1 XP_016884370.1 11 - 12 1902 87
XM_017028882.1 XP_016884371.1 11 - 12 1732 87
XM_017028883.1 XP_016884372.1 10 - 11 1395 87
XM_017028884.1 XP_016884373.1 12 - 13 2181 87
XM_017028887.1 XP_016884376.1 11 - 12 1732 87
GenBank mRNA AB037785.1 - 8 - 9 1054 87
AK127487.1 - 10 - 11 1631 87
BC157876.1 - 10 - 11 1611 87
BC171887.1 - 10 - 11 1611 87
BX647382.1 - 10 - 11 1572 87
CR456364.1 - 10 - 11 1564 87

View Full Product Details