The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006606.2 | NP_006597.2 | 2 - 3 | 219 | 81 |
GenBank mRNA | AF237576.1 | - | 2 - 3 | 173 | 81 |
AL832411.1 | - | 2 - 3 | 167 | 81 | |
BC015938.1 | - | 2 - 3 | 219 | 81 |