The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319310.1 | NP_001306239.1 | 3 - 4 | 695 | 144 |
NM_152317.3 | NP_689530.1 | 3 - 4 | 695 | 144 | |
NR_135073.1 | 3 - 4 | 695 | 144 | ||
XM_011537876.2 | XP_011536178.1 | 2 - 3 | 298 | 144 | |
XM_017018780.1 | XP_016874269.1 | 3 - 4 | 695 | 144 | |
XM_017018781.1 | XP_016874270.1 | 3 - 4 | 695 | 144 | |
GenBank mRNA | AK090824.1 | - | 3 - 4 | 695 | 144 |
BC015117.1 | - | 3 - 4 | 595 | 144 | |
BC100927.2 | - | 2 - 3 | 298 | 144 | |
BC100929.1 | - | 2 - 3 | 298 | 144 | |
BC100930.1 | - | 3 - 4 | 695 | 144 |