The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316967.1 | NP_001303896.1 | 5 - 6 | 931 | 107 |
NM_031302.4 | NP_112592.1 | 6 - 7 | 1066 | 107 | |
XM_011538793.2 | XP_011537095.1 | 6 - 7 | 1301 | 107 | |
XM_017019999.1 | XP_016875488.1 | 7 - 8 | 1382 | 107 | |
GenBank mRNA | AJ271711.1 | - | 3 - 4 | 280 | 107 |
AK027310.1 | - | 4 - 5 | 380 | 107 | |
AK056056.1 | - | 5 - 6 | 937 | 107 | |
AY358763.1 | - | 5 - 6 | 470 | 107 | |
BC022343.1 | - | 5 - 6 | 687 | 107 |