The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002205.4 | NP_002196.4 | 19 - 20 | 2051 | 54 |
GenBank mRNA | AK291570.1 | - | 19 - 20 | 1996 | 54 |
AK312414.1 | - | 19 - 20 | 2051 | 54 | |
BC008786.2 | - | 19 - 20 | 2036 | 54 | |
BX647780.1 | - | 8 - 9 | 1794 | 54 | |
M13918.2 | - | 19 - 20 | 1938 | 54 | |
X06256.1 | - | 19 - 20 | 2006 | 54 |