The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002205.4 | NP_002196.4 | 24 - 25 | 2560 | 66 |
GenBank mRNA | AK291570.1 | - | 24 - 25 | 2505 | 66 |
AK312414.1 | - | 24 - 25 | 2560 | 66 | |
BC008786.2 | - | 24 - 25 | 2545 | 66 | |
BX647780.1 | - | 13 - 14 | 2303 | 66 | |
M13918.2 | - | 24 - 25 | 2447 | 66 | |
X06256.1 | - | 24 - 25 | 2515 | 66 |