The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002205.4 | NP_002196.4 | 26 - 27 | 2793 | 68 |
GenBank mRNA | AK291570.1 | - | 26 - 27 | 2738 | 68 |
AK312414.1 | - | 26 - 27 | 2793 | 68 | |
BC008786.2 | - | 26 - 27 | 2778 | 68 | |
M13918.2 | - | 26 - 27 | 2680 | 68 | |
X06256.1 | - | 26 - 27 | 2748 | 68 |