The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145319.1 | NP_001138791.1 | 11 - 12 | 1415 | 102 |
NM_001172312.1 | NP_001165783.1 | 11 - 12 | 1576 | 102 | |
NM_002670.2 | NP_002661.2 | 11 - 12 | 1431 | 102 | |
XM_006713660.3 | XP_006713723.1 | 12 - 13 | 1726 | 102 | |
XM_011512900.2 | XP_011511202.1 | 13 - 14 | 1848 | 102 | |
XM_011512901.1 | XP_011511203.1 | 12 - 13 | 1655 | 102 | |
XM_011512903.2 | XP_011511205.1 | 12 - 13 | 1493 | 102 | |
XM_017006626.1 | XP_016862115.1 | 13 - 14 | 1898 | 102 | |
XM_017006627.1 | XP_016862116.1 | 12 - 13 | 1544 | 102 | |
GenBank mRNA | AK290316.1 | - | 11 - 12 | 1388 | 102 |
BC031083.1 | - | 11 - 12 | 1576 | 102 | |
L20826.1 | - | 11 - 12 | 1353 | 102 |