Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLS1
Entrez Gene ID
5357
Gene Name
plastin 1
Gene Aliases
-
Chromosome Location
Chr.3: 142596387 - 142713664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145319.1 NP_001138791.1 14 - 15 1785 138
NM_001172312.1 NP_001165783.1 14 - 15 1946 138
NM_002670.2 NP_002661.2 14 - 15 1801 138
XM_006713660.3 XP_006713723.1 15 - 16 2096 138
XM_011512900.2 XP_011511202.1 16 - 17 2218 138
XM_011512901.1 XP_011511203.1 15 - 16 2025 138
XM_011512903.2 XP_011511205.1 15 - 16 1863 138
XM_017006626.1 XP_016862115.1 16 - 17 2268 138
XM_017006627.1 XP_016862116.1 15 - 16 1914 138
GenBank mRNA AK290316.1 - 14 - 15 1758 138
BC031083.1 - 14 - 15 1946 138
L20826.1 - 14 - 15 1723 138

View Full Product Details