The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278439.1 | NP_001265368.1 | 14 - 15 | 1619 | 73 |
NM_007056.2 | NP_008987.2 | 15 - 16 | 1805 | 73 | |
XM_011526396.2 | XP_011524698.1 | 15 - 16 | 1800 | 73 | |
XM_011526397.2 | XP_011524699.1 | 14 - 15 | 1634 | 73 | |
GenBank mRNA | AF042800.1 | - | 15 - 16 | 1805 | 73 |
AK293333.1 | - | 14 - 15 | 1615 | 73 | |
BC013178.1 | - | 10 - 11 | 1091 | 73 | |
BC080554.1 | - | 14 - 15 | 1745 | 73 |