The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297555.1 | NP_001284484.1 | 4 - 5 | 546 | 63 |
NM_001297556.1 | NP_001284485.1 | 3 - 4 | 498 | 63 | |
NM_145185.3 | NP_660186.1 | 3 - 4 | 498 | 63 | |
XM_006722800.2 | XP_006722863.1 | 4 - 5 | 541 | 63 | |
GenBank mRNA | AF003199.1 | - | 3 - 4 | 394 | 63 |
AF006689.1 | - | 3 - 4 | 408 | 63 | |
AF013588.1 | - | 3 - 4 | 400 | 63 | |
AF014401.1 | - | 3 - 4 | 333 | 63 | |
AF022805.1 | - | 3 - 4 | 393 | 63 | |
AK300988.1 | - | 4 - 5 | 441 | 63 | |
AK313899.1 | - | 3 - 4 | 399 | 63 | |
BC038295.2 | - | 3 - 4 | 437 | 63 |