Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ANK2
- Entrez Gene ID
-
287
- Gene Name
- ankyrin 2, neuronal
- Gene Aliases
-
ANK-2, LQT4, brank-2
- Chromosome Location
-
Chr.4: 112706083 - 113383740 on Build GRCh38
View Full Product Details