The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002440.3 | NP_002431.2 | 10 - 11 | 1482 | 132 |
GenBank mRNA | AF104243.1 | - | 10 - 11 | 1378 | 132 |
BC033030.1 | - | 10 - 11 | 1472 | 132 | |
U89293.1 | - | 10 - 11 | 1408 | 132 |