Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSH4
Entrez Gene ID
4438
Gene Name
mutS homolog 4
Gene Aliases
-
Chromosome Location
Chr.1: 75796871 - 75913238 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002440.3 NP_002431.2 12 - 13 1786 177
GenBank mRNA AF104243.1 - 12 - 13 1682 177
BC033030.1 - 12 - 13 1776 177
U89293.1 - 12 - 13 1712 177

View Full Product Details