The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006012.2 | NP_006003.1 | 1 - 2 | 326 | 68 |
GenBank mRNA | AK311973.1 | - | 1 - 2 | 226 | 68 |
BC002956.1 | - | 1 - 2 | 326 | 68 | |
Z50853.1 | - | 1 - 2 | 222 | 68 |