The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016131.4 | NP_057215.3 | 1 - 2 | 873 | 111 |
GenBank mRNA | AF086917.1 | - | 1 - 2 | 218 | 111 |
AF106681.1 | - | 1 - 2 | 214 | 111 | |
AF297660.1 | - | 1 - 2 | 124 | 111 | |
AF498945.1 | - | 1 - 2 | 124 | 111 | |
AK023223.1 | - | 1 - 2 | 623 | 111 | |
AL136650.1 | - | 1 - 2 | 632 | 111 | |
AL517754.3 | - | 1 - 2 | 634 | 111 | |
BC000896.1 | - | 1 - 2 | 199 | 111 | |
CR457303.1 | - | 1 - 2 | 124 | 111 |