The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000182.4 | NP_000173.2 | 11 - 12 | 1216 | 81 |
GenBank mRNA | AK293398.1 | - | 8 - 9 | 709 | 81 |
AK299258.1 | - | 11 - 12 | 964 | 81 | |
AK313027.1 | - | 11 - 12 | 1109 | 81 | |
BC009235.2 | - | 11 - 12 | 1111 | 81 | |
D16480.1 | - | 11 - 12 | 1113 | 81 | |
FJ626228.1 | - | 10 - 11 | 952 | 81 | |
GU727642.1 | - | 11 - 12 | 1111 | 81 | |
U04627.1 | - | 11 - 12 | 1112 | 81 |