The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006610.3 | NP_006601.2 | 7 - 8 | 1041 | 108 |
GenBank mRNA | AK290823.1 | - | 7 - 8 | 1041 | 108 |
X98400.1 | - | 7 - 8 | 1045 | 108 | |
Y09926.1 | - | 7 - 8 | 1025 | 108 |