The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006610.3 | NP_006601.2 | 8 - 9 | 1120 | 120 |
GenBank mRNA | AK290823.1 | - | 8 - 9 | 1120 | 120 |
X98400.1 | - | 8 - 9 | 1124 | 120 | |
Y09926.1 | - | 8 - 9 | 1104 | 120 |