The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006610.3 | NP_006601.2 | 9 - 10 | 1253 | 88 |
GenBank mRNA | X98400.1 | - | 9 - 10 | 1257 | 88 |
Y09926.1 | - | 9 - 10 | 1237 | 88 |