Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLL2
Entrez Gene ID
7093
Gene Name
tolloid like 2
Gene Aliases
-
Chromosome Location
Chr.10: 96364606 - 96513926 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012465.3 NP_036597.1 14 - 15 2095 114
GenBank mRNA AB023149.2 - 14 - 15 2045 114
AF059516.1 - 14 - 15 2218 114
BC112341.1 - 14 - 15 1978 114
BC112366.1 - 14 - 15 1978 114
BC113577.1 - 14 - 15 1911 114

View Full Product Details