Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLL2
Entrez Gene ID
7093
Gene Name
tolloid like 2
Gene Aliases
-
Chromosome Location
Chr.10: 96364606 - 96513926 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012465.3 NP_036597.1 9 - 10 1407 86
GenBank mRNA AB023149.2 - 9 - 10 1357 86
AF059516.1 - 9 - 10 1530 86
AK026106.1 - 9 - 10 1333 86
BC013871.2 - 9 - 10 1367 86
BC112341.1 - 9 - 10 1290 86
BC112366.1 - 9 - 10 1290 86
BC113577.1 - 9 - 10 1223 86

View Full Product Details