The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143682.1 | NP_001137154.1 | 10 - 11 | 1369 | 70 |
NM_020898.2 | NP_065949.1 | 11 - 12 | 1624 | 70 | |
NR_026554.1 | 10 - 11 | 1521 | 70 | ||
XM_011538601.1 | XP_011536903.1 | 11 - 12 | 1558 | 70 | |
XM_011538602.1 | XP_011536904.1 | 12 - 13 | 1691 | 70 | |
XM_011538603.1 | XP_011536905.1 | 12 - 13 | 1709 | 70 | |
XM_011538604.1 | XP_011536906.1 | 12 - 13 | 1665 | 70 | |
XM_017019707.1 | XP_016875196.1 | 12 - 13 | 1691 | 70 | |
XM_017019708.1 | XP_016875197.1 | 10 - 11 | 1686 | 70 | |
XM_017019709.1 | XP_016875198.1 | 10 - 11 | 1686 | 70 | |
GenBank mRNA | AB040969.1 | - | 11 - 12 | 1521 | 70 |
AF370415.1 | - | 11 - 12 | 1529 | 70 | |
AK027881.1 | - | 11 - 12 | 1558 | 70 | |
AK122773.1 | - | 10 - 11 | 1285 | 70 | |
AK294424.1 | - | 11 - 12 | 1532 | 70 | |
AK315578.1 | - | 11 - 12 | 1559 | 70 | |
AL136895.1 | - | 11 - 12 | 1536 | 70 | |
AY358397.1 | - | 11 - 12 | 1533 | 70 | |
AY563137.1 | - | 11 - 12 | 1531 | 70 | |
BC003177.1 | - | 11 - 12 | 1531 | 70 |