Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNPR
Entrez Gene ID
132204
Gene Name
synaptoporin
Gene Aliases
SPO
Chromosome Location
Chr.3: 63200604 - 63616924 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130003.1 NP_001123475.1 2 - 3 380 69
XM_017005732.1 XP_016861221.1 2 - 3 380 69
GenBank mRNA AK293628.1 - 2 - 3 291 69
CR936762.1 - 2 - 3 301 69

View Full Product Details