Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNPR
Entrez Gene ID
132204
Gene Name
synaptoporin
Gene Aliases
SPO
Chromosome Location
Chr.3: 63200604 - 63616924 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130003.1 NP_001123475.1 4 - 5 701 80
NM_144642.4 NP_653243.1 3 - 4 714 80
XM_017005731.1 XP_016861220.1 4 - 5 467 80
XM_017005732.1 XP_016861221.1 4 - 5 701 80
GenBank mRNA AF411860.2 - 3 - 4 463 80
AK122831.1 - 4 - 5 491 80
AK309017.1 - 3 - 4 496 80
AK312465.1 - 3 - 4 619 80
AL834457.1 - 3 - 4 443 80
BC022518.1 - 3 - 4 463 80
CR936762.1 - 4 - 5 622 80

View Full Product Details