The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032228.5 | NP_115604.1 | 1 - 2 | 150 | 141 |
XM_011520400.1 | XP_011518702.1 | 1 - 2 | 150 | 141 | |
GenBank mRNA | AY358784.1 | - | 1 - 2 | 139 | 141 |
CR936619.1 | - | 1 - 2 | 125 | 141 | |
DA515509.1 | - | 1 - 2 | 150 | 141 |