Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HR
Entrez Gene ID
55806
Gene Name
hair growth associated
Gene Aliases
ALUNC, AU, HSA277165, HYPT4, MUHH, MUHH1
Chromosome Location
Chr.8: 22114419 - 22131053 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005144.4 NP_005135.2 14 - 15 3642 140
NM_018411.4 NP_060881.2 14 - 15 3642 140
XM_005273569.1 XP_005273626.1 14 - 15 3646 140
XM_006716367.1 XP_006716430.1 14 - 15 3646 140
GenBank mRNA AF039196.3 - 14 - 15 4461 140
AJ277165.2 - 13 - 14 3017 140
BC067128.1 - 14 - 15 3107 140

View Full Product Details