Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HR
Entrez Gene ID
55806
Gene Name
hair growth associated
Gene Aliases
ALUNC, AU, HSA277165, HYPT4, MUHH, MUHH1
Chromosome Location
Chr.8: 22114419 - 22131053 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005144.4 NP_005135.2 7 - 8 2668 69
NM_018411.4 NP_060881.2 7 - 8 2668 69
XM_005273569.1 XP_005273626.1 7 - 8 2672 69
XM_006716367.1 XP_006716430.1 7 - 8 2672 69
GenBank mRNA AF039196.3 - 7 - 8 3487 69
AJ277165.2 - 6 - 7 2043 69
BC067128.1 - 7 - 8 2133 69

View Full Product Details