Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIAS1
Entrez Gene ID
8554
Gene Name
protein inhibitor of activated STAT 1
Gene Aliases
DDXBP1, GBP, GU/RH-II, ZMIZ3
Chromosome Location
Chr.15: 68054179 - 68191464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320687.1 NP_001307616.1 11 - 12 2047 99
NM_016166.2 NP_057250.1 10 - 11 1448 99
XM_011522126.2 XP_011520428.1 12 - 13 2155 99
XM_011522127.2 XP_011520429.1 10 - 11 1932 99
XM_017022688.1 XP_016878177.1 10 - 11 1926 99
XM_017022689.1 XP_016878178.1 10 - 11 1939 99
XM_017022690.1 XP_016878179.1 10 - 11 1744 99
XM_017022691.1 XP_016878180.1 10 - 11 1518 99
GenBank mRNA AF077951.1 - 10 - 11 1297 99
AF167160.1 - 10 - 11 1396 99
AK094641.1 - 11 - 12 2047 99
AK314515.1 - 10 - 11 1323 99
AY826819.1 - 10 - 11 1297 99
BC053685.1 - 10 - 11 1301 99
BC118587.1 - 10 - 11 1350 99
BC121797.1 - 10 - 11 1347 99
EF487997.1 - 10 - 11 1300 99
EF487998.1 - 10 - 11 1300 99
U78524.1 - 9 - 10 1282 99

View Full Product Details