Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIAS1
Entrez Gene ID
8554
Gene Name
protein inhibitor of activated STAT 1
Gene Aliases
DDXBP1, GBP, GU/RH-II, ZMIZ3
Chromosome Location
Chr.15: 68054179 - 68191464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320687.1 NP_001307616.1 12 - 13 2232 72
NM_016166.2 NP_057250.1 11 - 12 1633 72
XM_011522126.2 XP_011520428.1 13 - 14 2340 72
XM_011522127.2 XP_011520429.1 11 - 12 2117 72
XM_017022688.1 XP_016878177.1 11 - 12 2111 72
XM_017022689.1 XP_016878178.1 11 - 12 2124 72
XM_017022690.1 XP_016878179.1 11 - 12 1929 72
XM_017022691.1 XP_016878180.1 11 - 12 1703 72
GenBank mRNA AF077951.1 - 11 - 12 1482 72
AF167160.1 - 11 - 12 1581 72
AK094641.1 - 12 - 13 2232 72
AK314515.1 - 11 - 12 1508 72
AY826819.1 - 11 - 12 1482 72
BC118587.1 - 11 - 12 1535 72
BC121797.1 - 11 - 12 1532 72
EF487998.1 - 11 - 12 1485 72
U78524.1 - 10 - 11 1467 72

View Full Product Details