Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIAS1
Entrez Gene ID
8554
Gene Name
protein inhibitor of activated STAT 1
Gene Aliases
DDXBP1, GBP, GU/RH-II, ZMIZ3
Chromosome Location
Chr.15: 68054179 - 68191464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320687.1 NP_001307616.1 4 - 5 1298 78
NM_016166.2 NP_057250.1 3 - 4 699 78
XM_011522126.2 XP_011520428.1 5 - 6 1406 78
XM_011522127.2 XP_011520429.1 3 - 4 1183 78
XM_017022688.1 XP_016878177.1 3 - 4 1177 78
XM_017022689.1 XP_016878178.1 3 - 4 1190 78
XM_017022690.1 XP_016878179.1 3 - 4 995 78
XM_017022691.1 XP_016878180.1 3 - 4 769 78
GenBank mRNA AF077951.1 - 3 - 4 551 78
AF167160.1 - 3 - 4 647 78
AK094641.1 - 4 - 5 1298 78
AK314515.1 - 3 - 4 574 78
AY826819.1 - 3 - 4 548 78
BC053685.1 - 3 - 4 552 78
BC104646.1 - 3 - 4 567 78
BC118587.1 - 3 - 4 601 78
BC121797.1 - 3 - 4 598 78
EF487997.1 - 3 - 4 551 78
EF487998.1 - 3 - 4 551 78
FJ997900.1 - 3 - 4 551 78
U78524.1 - 2 - 3 533 78

View Full Product Details