Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCNN1B
Entrez Gene ID
6338
Gene Name
sodium channel epithelial 1 beta subunit
Gene Aliases
BESC1, ENaCb, ENaCbeta, SCNEB
Chromosome Location
Chr.16: 23278231 - 23381299 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000336.2 NP_000327.2 1 - 2 168 142
GenBank mRNA BC036352.1 - 1 - 2 127 142
X87159.1 - 1 - 2 119 142

View Full Product Details