Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS10
Entrez Gene ID
81794
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 10
Gene Aliases
ADAM-TS10, ADAMTS-10, WMS, WMS1
Chromosome Location
Chr.19: 8580240 - 8610735 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030957.3 NP_112219.3 12 - 13 1782 82
XM_006722917.3 XP_006722980.1 11 - 12 1628 82
XM_017027338.1 XP_016882827.1 11 - 12 1638 82
GenBank mRNA AB209515.1 - 11 - 12 1618 82
AF163762.1 - 9 - 10 1398 82
BM928283.1 - - 539 82

View Full Product Details