Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTS10
Entrez Gene ID
81794
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 10
Gene Aliases
ADAM-TS10, ADAMTS-10, WMS, WMS1
Chromosome Location
Chr.19: 8580240 - 8610735 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030957.3 NP_112219.3 16 - 17 2207 101
XM_006722917.3 XP_006722980.1 15 - 16 2053 101
XM_017027338.1 XP_016882827.1 15 - 16 2063 101
XM_017027339.1 XP_016882828.1 6 - 7 687 101
XM_017027340.1 XP_016882829.1 5 - 6 547 101
GenBank mRNA AB209515.1 - 15 - 16 2043 101
AF163762.1 - 13 - 14 1823 101

View Full Product Details