Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTS10
Entrez Gene ID
81794
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 10
Gene Aliases
ADAM-TS10, ADAMTS-10, WMS, WMS1
Chromosome Location
Chr.19: 8580240 - 8610735 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282352.1 NP_001269281.1 7 - 8 895 128
NM_030957.3 NP_112219.3 20 - 21 2706 128
XM_006722917.3 XP_006722980.1 19 - 20 2552 128
XM_017027338.1 XP_016882827.1 19 - 20 2562 128
XM_017027339.1 XP_016882828.1 10 - 11 1186 128
XM_017027340.1 XP_016882829.1 9 - 10 1046 128
GenBank mRNA AB209515.1 - 19 - 20 2542 128
AF163762.1 - 17 - 18 2322 128
AK131414.1 - 7 - 8 895 128
BC029308.1 - 1 - 2 53 128

View Full Product Details