The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030957.3 | NP_112219.3 | 2 - 3 | 212 | 64 |
GenBank mRNA | BI822988.1 | - | 2 - 3 | 152 | 64 |
CD632934.1 | - | 2 - 3 | 175 | 64 | |
CD632956.1 | - | 2 - 3 | 147 | 64 |