The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005309.2 | NP_005300.1 | 1 - 2 | 381 | 64 |
XM_011516993.2 | XP_011515295.1 | 2 - 3 | 1071 | 64 | |
GenBank mRNA | AK225562.1 | - | 1 - 2 | 381 | 64 |
AK298488.1 | - | 1 - 2 | 339 | 64 | |
AK303779.1 | - | 1 - 2 | 323 | 64 | |
BC018207.1 | - | 2 - 3 | 315 | 64 | |
BP275218.1 | - | 1 - 2 | 381 | 64 | |
BT006992.1 | - | 1 - 2 | 158 | 64 | |
D10355.1 | - | 1 - 2 | 149 | 64 |