The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005309.2 | NP_005300.1 | 4 - 5 | 714 | 71 |
XM_011516993.2 | XP_011515295.1 | 5 - 6 | 1404 | 71 | |
GenBank mRNA | AK225562.1 | - | 4 - 5 | 714 | 71 |
AK298488.1 | - | 4 - 5 | 672 | 71 | |
AK303779.1 | - | 3 - 4 | 774 | 71 | |
BC018207.1 | - | 5 - 6 | 648 | 71 | |
BT006992.1 | - | 4 - 5 | 491 | 71 | |
D10355.1 | - | 4 - 5 | 482 | 71 |